Boc-(S)-3-amino-4-(4-nitrophenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(S)-3-((tert-Butoxycarbonyl)amino)-4-(4-nitrophenyl)butanoic acid
042261-250MG
042261-1G
101855-110EA
275.38
USD
101855-110
101855-112
Boc-(S)-3-amino-4-(4-nitrophenyl)butyric acid
Boc-(S)-3-amino-4-(4-nitrophenyl)butyric acid
Hazard: Irritant
Formula:
C₁₅H₂₀N₂O₆ MW: 324.34 g/mol |
MDL Number:
MFCD01076286 CAS Number: 127106-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...