Boc-(S)-3-amino-4-(4-methylphenyl)butyric acid

Supplier: Matrix Scientific

042260-250MG 042260-1G
101855-106EA 275.38 USD
101855-106 101855-108
Boc-(S)-3-amino-4-(4-methylphenyl)butyric acid
Boc-(S)-3-amino-4-(4-methylphenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₃NO₄
MDL Number: MFCD01861026
CAS Number: 270062-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR