Boc-(S)-3-amino-4-(4-fluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(4-fluorophenyl)butanoic acid

042258-250MG 042258-1G
101855-098EA 275.38 USD
101855-098 101855-100
Boc-(S)-3-amino-4-(4-fluorophenyl)butyric acid
Boc-(S)-3-amino-4-(4-fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₂₀FNO₄
MW: 297.33 g/mol
MDL Number: MFCD01076277
CAS Number: 218608-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR