Boc-(S)-3-amino-4-(4-cyanophenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(S)-3-((tert-Butoxycarbonyl)amino)-4-(4-cyanophenyl)butanoic acid
042257-250MG
042257-1G
101855-094EA
373.85
USD
101855-094
101855-096
Boc-(S)-3-amino-4-(4-cyanophenyl)butyric acid
Boc-(S)-3-amino-4-(4-cyanophenyl)butyric acid
Hazard: Irritant
Formula:
C₁₆H₂₀N₂O₄ MW: 304.35 g/mol |
MDL Number:
MFCD01861079 CAS Number: 270065-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...