Boc-(S)-3-amino-4-(4-chlorophenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(S)-3-((tert-Butoxycarbonyl)amino)-4-(4-chlorophenyl)butanoic acid
042256-250MG
042256-1G
101855-090EA
275.38
USD
101855-090
101855-092
Boc-(S)-3-amino-4-(4-chlorophenyl)butyric acid
Boc-(S)-3-amino-4-(4-chlorophenyl)butyric acid
Hazard: Irritant
Formula:
C₁₅H₂₀ClNO₄ MW: 313.78 g/mol |
MDL Number:
MFCD01318372 CAS Number: 270596-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...