Boc-(S)-3-amino-4-(3-pyridyl)butyric acid

Supplier: Matrix Scientific

042252-250MG 042252-1G
101855-074EA 510.77 USD
101855-074 101855-076
Boc-(S)-3-amino-4-(3-pyridyl)butyric acid
Boc-(S)-3-amino-4-(3-pyridyl)butyric acid

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₄
MW: 280.33 g/mol
MDL Number: MFCD01861056
CAS Number: 208404-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR