Boc-(S)-3-amino-4-(3-fluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(3-fluorophenyl)butanoic acid

042250-250MG 042250-1G
101855-066EA 275.38 USD
101855-066 101855-068
Boc-(S)-3-amino-4-(3-fluorophenyl)butyric acid
Boc-(S)-3-amino-4-(3-fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₂₀FNO₄
MW: 297.33 g/mol
MDL Number: MFCD01861011
CAS Number: 270596-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR