Boc-(S)-3-amino-4-(3-cyanophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(3-cyanophenyl)butanoic acid

042249-250MG 042249-1G
101855-062EA 373.85 USD
101855-062 101855-064
Boc-(S)-3-amino-4-(3-cyanophenyl)butyric acid
Boc-(S)-3-amino-4-(3-cyanophenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₄
MW: 304.35 g/mol
MDL Number: MFCD01861076
CAS Number: 270065-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR