Boc-(S)-3-amino-4-(3-benzothienyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-4-(Benzo[b]thiophen-3-yl)-3-((tert-butoxycarbonyl)amino)butanoic acid

042247-250MG 042247-1G
101855-054EA 349.23 USD
101855-054 101855-056
Boc-(S)-3-amino-4-(3-benzothienyl)butyric acid
Boc-(S)-3-amino-4-(3-benzothienyl)butyric acid

Hazard: Irritant

Formula: C₁₇H₂₁NO₄S
MW: 335.43 g/mol
MDL Number: MFCD01861041
CAS Number: 270063-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR