Boc-(S)-3-amino-4-(3,4-dichlorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(3,4-dichlorophenyl)butanoic acid

042245-250MG 042245-1G
101855-046EA 113.85 USD
101855-046 101855-048
Boc-(S)-3-amino-4-(3,4-dichlorophenyl)butyric acid
Boc-(S)-3-amino-4-(3,4-dichlorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₁₉Cl₂NO₄
MW: 348.23 g/mol
MDL Number: MFCD01861047
CAS Number: 270063-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR