Boc-(S)-3-amino-4-(2-trifluoromethylphenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(2-(trifluoromethyl)phenyl)butanoic acid

042244-250MG 042244-1G
101855-042EA 412.31 USD
101855-042 101855-044
Boc-(S)-3-amino-4-(2-trifluoromethylphenyl)butyric acid
Boc-(S)-3-amino-4-(2-trifluoromethylphenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₀F₃NO₄
MW: 347.34 g/mol
MDL Number: MFCD01861064
CAS Number: 270065-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR