Boc-(S)-3-amino-4-(2-naphthyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(naphthalen-2-yl)butanoic acid

042242-250MG 042242-1G
101855-034EA 275.38 USD
101855-034 101855-036
Boc-(S)-3-amino-4-(2-naphthyl)butyric acid
Boc-(S)-3-amino-4-(2-naphthyl)butyric acid

Hazard: Irritant

Formula: C₁₉H₂₃NO₄
MW: 329.4 g/mol
MDL Number: MFCD01076284
CAS Number: 219297-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR