Boc-(S)-3-amino-4-(2-furyl)butyric acid

Supplier: Matrix Scientific

042240-250MG 042240-1G
101855-026EA 498.46 USD
101855-026 101855-028
Boc-(S)-3-amino-4-(2-furyl)butyric acid
Boc-(S)-3-amino-4-(2-furyl)butyric acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₅
MW: 269.3 g/mol
MDL Number: MFCD01861090
CAS Number: 270263-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR