Boc-(S)-3-amino-4-(2-cyanophenyl)butyric acid

Supplier: Matrix Scientific

042238-250MG 042238-1G
101855-018EA 373.85 USD
101855-018 101855-020
Boc-(S)-3-amino-4-(2-cyanophenyl)butyric acid
Boc-(S)-3-amino-4-(2-cyanophenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₄
MDL Number: MFCD01861073
CAS Number: 270065-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR