Boc-(S)-3-amino-4-(2,4-dichlorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(2,4-dichlorophenyl)butanoic acid

042236-250MG 042236-1G
101855-010EA 349.23 USD
101855-010 101855-012
Boc-(S)-3-amino-4-(2,4-dichlorophenyl)butyric acid
Boc-(S)-3-amino-4-(2,4-dichlorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₁₉Cl₂NO₄
MDL Number: MFCD01861044
CAS Number: 270063-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR