Boc-(R)-3-amino-6-phenyl-5-hexenoic acid
Supplier: Matrix Scientific
042232-250MG
042232-1G
101854-994EA
412.31
USD
101854-994
101854-996
Boc-(R)-3-amino-6-phenyl-5-hexenoic acid
Boc-(R)-3-amino-6-phenyl-5-hexenoic acid
Hazard: Irritant
Formula:
C₁₇H₂₃NO₄ |
MDL Number:
MFCD01860906 CAS Number: 332064-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...