Boc-(R)-3-amino-5-phenylpentanoic acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((tert-Butoxycarbonyl)amino)-5-phenylpentanoic acid
042231-250MG
042231-1G
101854-990EA
275.38
USD
101854-990
101854-992
Boc-(R)-3-amino-5-phenylpentanoic acid
Boc-(R)-3-amino-5-phenylpentanoic acid
Hazard: Irritant
Formula:
C₁₆H₂₃NO₄ MW: 293.37 g/mol |
MDL Number:
MFCD01076262 CAS Number: 218608-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...