Boc-(R)-3-amino-5-hexynoic acid

Supplier: Matrix Scientific

042230-250MG 042230-1G
101854-986EA 324.62 USD
101854-986 101854-988
Boc-(R)-3-amino-5-hexynoic acid
Boc-(R)-3-amino-5-hexynoic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD01860909
CAS Number: 332064-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR