Boc-(R)-3-amino-5-hexenoic acid
Supplier: Matrix Scientific
Synonyms:
Boc-(R)-3-Amino-5-hexenoic acid, Boc-D-β-HoGly(Allyl)-OH
042229-250MG
042229-1G
101854-982EA
412.31
USD
101854-982
101854-984
Boc-(R)-3-amino-5-hexenoic acid
Boc-(R)-3-amino-5-hexenoic acid
Hazard: Irritant
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol |
MDL Number:
MFCD01860990 CAS Number: 269726-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...