Boc-(R)-3-amino-4-(4-trifluoromethylphenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(4-(trifluoromethyl)phenyl)butanoic acid

042226-250MG 042226-1G
101854-970EA 412.31 USD
101854-970 101854-972
Boc-(R)-3-amino-4-(4-trifluoromethylphenyl)butyric acid
Boc-(R)-3-amino-4-(4-trifluoromethylphenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₀F₃NO₄
MW: 347.34 g/mol
MDL Number: MFCD01860972
CAS Number: 269726-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR