Boc-(R)-3-amino-4-(4-methylphenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(p-tolyl)butanoic acid

042223-250MG 042223-1G
101854-958EA 275.38 USD
101854-958 101854-960
Boc-(R)-3-amino-4-(4-methylphenyl)butyric acid
Boc-(R)-3-amino-4-(4-methylphenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₃NO₄
MW: 293.37 g/mol
MDL Number: MFCD01860928
CAS Number: 269398-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR