Boc-(R)-3-amino-4-(4-iodophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(4-iodophenyl)butanoic acid

042222-250MG 042222-1G
101854-954EA 275.38 USD
101854-954 101854-956
Boc-(R)-3-amino-4-(4-iodophenyl)butyric acid
Boc-(R)-3-amino-4-(4-iodophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₂₀INO₄
MW: 405.24 g/mol
MDL Number: MFCD01860963
CAS Number: 269396-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR