Boc-(R)-3-amino-4-(4-fluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(4-fluorophenyl)butanoic acid

042221-250MG 042221-1G
101854-950EA 275.38 USD
101854-950 101854-952
Boc-(R)-3-amino-4-(4-fluorophenyl)butyric acid
Boc-(R)-3-amino-4-(4-fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₂₀FNO₄
MW: 297.33 g/mol
MDL Number: MFCD01076280
CAS Number: 218609-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR