Boc-(R)-3-amino-4-(4-chlorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(4-chlorophenyl)butanoic acid

042219-250MG 042219-1G
101854-942EA 193.81 USD
101854-942 101854-944
Boc-(R)-3-amino-4-(4-chlorophenyl)butyric acid
Boc-(R)-3-amino-4-(4-chlorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₂₀ClNO₄
MW: 313.78 g/mol
MDL Number: MFCD01076276
CAS Number: 218608-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR