Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-4-(4-Bromophenyl)-3-((tert-butoxycarbonyl)amino)butanoic acid

042218-1G 042218-5G
101854-938EA 823.08 USD
101854-938 101854-940
Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid
Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₂₀BrNO₄
MW: 358.24 g/mol
MDL Number: MFCD01860917
CAS Number: 331763-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR