Boc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(3-(trifluoromethyl)phenyl)butanoic acid

042217-250MG 042217-1G
101854-934EA 412.31 USD
101854-934 101854-936
Boc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid
Boc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₀F₃NO₄
MW: 347.34 g/mol
MDL Number: MFCD01860969
CAS Number: 269726-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR