Boc-(R)-3-amino-4-(3-pyridyl)butyric acid

Supplier: Matrix Scientific

042215-250MG 042215-1G
101854-926EA 510.77 USD
101854-926 101854-928
Boc-(R)-3-amino-4-(3-pyridyl)butyric acid
Boc-(R)-3-amino-4-(3-pyridyl)butyric acid

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₄
MW: 280.33 g/mol
MDL Number: MFCD01860958
CAS Number: 269396-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR