Boc-(R)-3-amino-4-(3-methylphenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((tert-Butoxycarbonyl)amino)-4-(m-tolyl)butanoic acid
042214-250MG
042214-1G
101854-922EA
275.38
USD
101854-922
101854-924
Boc-(R)-3-amino-4-(3-methylphenyl)butyric acid
Boc-(R)-3-amino-4-(3-methylphenyl)butyric acid
Hazard: Irritant
Formula:
C₁₆H₂₃NO₄ MW: 293.37 g/mol |
MDL Number:
MFCD01860925 CAS Number: 269398-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...