Boc-(R)-3-amino-4-(3-cyanophenyl)butyric acid

Supplier: Matrix Scientific

042212-250MG 042212-1G
101854-914EA 373.85 USD
101854-914 101854-916
Boc-(R)-3-amino-4-(3-cyanophenyl)butyric acid
Boc-(R)-3-amino-4-(3-cyanophenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₄
MDL Number: MFCD01860978
CAS Number: 269726-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR