Boc-(R)-3-amino-4-(3-benzothienyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-4-(Benzo[b]thiophen-3-yl)-3-((tert-butoxycarbonyl)amino)butanoic acid

042210-250MG 042210-1G
101854-906EA 349.23 USD
101854-906 101854-908
Boc-(R)-3-amino-4-(3-benzothienyl)butyric acid
Boc-(R)-3-amino-4-(3-benzothienyl)butyric acid

Hazard: Irritant

Formula: C₁₇H₂₁NO₄S
MW: 335.43 g/mol
MDL Number: MFCD01860943
CAS Number: 190190-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR