Boc-(R)-3-amino-4-(3,4-difluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(3,4-difluorophenyl)butanoic acid

042209-250MG 042209-1G
101854-902EA 235.38 USD
101854-902 101854-904
Boc-(R)-3-amino-4-(3,4-difluorophenyl)butyric acid
Boc-(R)-3-amino-4-(3,4-difluorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₁₉F₂NO₄
MW: 315.32 g/mol
MDL Number: MFCD01860952
CAS Number: 269396-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR