Boc-(R)-3-amino-4-(2-trifluoromethylphenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2-(trifluoromethyl)phenyl)butanoic acid
042207-250MG
042207-1G
101854-894EA
412.31
USD
101854-894
101854-896
Boc-(R)-3-amino-4-(2-trifluoromethylphenyl)butyric acid
Boc-(R)-3-amino-4-(2-trifluoromethylphenyl)butyric acid
Hazard: Irritant
Formula:
C₁₆H₂₀F₃NO₄ MW: 347.34 g/mol |
MDL Number:
MFCD01860966 CAS Number: 269396-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...