Methyl piperidin-4-ylacetate

Supplier: Matrix Scientific

039965-1G 039965-5G 039965-25G
101848-558EA 60 USD
101848-558 101848-560 101848-562
Methyl piperidin-4-ylacetate
Methyl piperidin-4-ylacetate

Hazard: Irritant

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD04115353
CAS Number: 168986-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR