2-(tert-Butoxycarbonylamino)-1-ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonyl)ethanolamine, N-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(Boc-amino)-1-ethanol, Boc-Glycinol

039950-25G 039950-100G
101848-524EA 23.08 USD
101848-524 101848-526
2-(tert-Butoxycarbonylamino)-1-ethanol ≥95%
2-(tert-Butoxycarbonylamino)-1-ethanol

Hazard: Irritant/Refrigerate

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Boiling Pt: 92 °C (.22 mmHg)
Density: 1.042
MDL Number: MFCD00056657
CAS Number: 26690-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR