5-(Aminomethyl)-2-chloropyridine

Supplier: Matrix Scientific

Synonyms: (6-Chloropyridin-3-yl)methanamine

039942-5G 039942-25G
101848-504EA 126.15 USD
101848-504 101848-506
5-(Aminomethyl)-2-chloropyridine
5-(Aminomethyl)-2-chloropyridine

Hazard: Irritant

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD00673153
CAS Number: 97004-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR