4,5-Dimethoxyisatoic anhydride
Supplier: Matrix Scientific
Synonyms:
6,7-Dimethoxy-1H-benzo[d][1,3]oxazine-2,4-dione
039854-1G
039854-5G
101848-324EA
60
USD
101848-324
101848-326
4,5-Dimethoxyisatoic anhydride
4,5-Dimethoxyisatoic anhydride
Hazard: Irritant
Formula:
C₁₀H₉NO₅ MW: 223.1800 g/mol |
MDL Number:
MFCD03426390 CAS Number: 20197-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...