2-Amino-3-chloropyrazine ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Chloropyrazin-2-amine, 3-Chloro-2-pyrazinamine
039812-1G
039812-5G
039812-25G
101848-236EA
181.54
USD
101848-236
101848-238
101848-240
2-Amino-3-chloropyrazine ≥97%
2-Amino-3-chloropyrazine
Hazard: Irritant
Formula:
C₄H₄ClN₃ MW: 129.55 g/mol |
MDL Number:
MFCD04114305 CAS Number: 6863-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...