2-(1-Cyclohexenyl)cyclohexanone

Supplier: Matrix Scientific

Synonyms: [1,1'-Bi(cyclohexan)]-1'-en-2-one

037869-5G 037869-100G
101844-344EA 26.15 USD
101844-344 101844-348
2-(1-Cyclohexenyl)cyclohexanone
2-(1-Cyclohexenyl)cyclohexanone

Hazard: Irritant

Formula: C₁₂H₁₈O
MW: 178.28 g/mol
MDL Number: MFCD00013796
CAS Number: 1502-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR