1-Boc-isonipecotic acid methyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl piperidine-4-carboxylate, N-BOC protected, 1-tert-Butyl 4-methyl piperidine-1,4-dicarboxylate

037710-5G 037710-100G
101844-002EA 15.38 USD
101844-002 101844-006
1-Boc-isonipecotic acid methyl ester ≥95%
1-Boc-isonipecotic acid methyl ester

Hazard: Irritant/Store in Freezer

Formula: C₁₂H₂₁NO₄
MW: 243.2994 g/mol
Melting Pt: 35 °C
MDL Number: MFCD02183584
CAS Number: 124443-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR