1-Boc-4-(aminomethyl)piperidine

Supplier: Matrix Scientific

Synonyms: 4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate, tert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate, 4-(Aminomethyl)-1-Boc-piperidine, 4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-4-(Aminomethyl)piperidine

037605-1G 037605-25G
101843-772EA 606.15 USD
101843-772 101843-776
1-Boc-4-(aminomethyl)piperidine
1-Boc-4-(aminomethyl)piperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 237…238 °C
Density: 1.013
MDL Number: MFCD01076207
CAS Number: 144222-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR