4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol ≥95%

Supplier: Matrix Scientific

037592-500MG 037592-5G
101843-734EA 116.92 USD
101843-734 101843-738
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol ≥95%
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol

Hazard: Irritant

Formula: C₈H₁₁NO₃
MDL Number: MFCD00025592
CAS Number: 51-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR