3,4-Diaminoanisole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxy-o-phenylenediamine

037533-5G 037533-25G
101843-608EA 123.08 USD
101843-608 101843-610
3,4-Diaminoanisole ≥95%
3,4-Diaminoanisole

Hazard: Toxic/Light Sensitive/Air Sensitive

Formula: C₇H₁₀N₂O
Melting Pt: 46…50 °C
MDL Number: MFCD00047837
CAS Number: 102-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR