2,5-Dimethylphenylhydrazinium chloride

Supplier: Matrix Scientific

Synonyms: 1-(2,5-Dimethylphenyl)hydrazine hydrochloride, 2,5-Dimethylphenylhydrazine hydrochloride

037512-5G 037512-25G
101843-568EA 43.08 USD
101843-568 101843-570
2,5-Dimethylphenylhydrazinium chloride
2,5-Dimethylphenylhydrazinium chloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂
MW: 172.66
MDL Number: MFCD00013382
CAS Number: 56737-78-1
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR