(R)-(-)-Thiazolidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid

037460-5G 037460-25G
101843-440EA 27.69 USD
101843-440 101843-442
(R)-(-)-Thiazolidine-4-carboxylic acid ≥95%
(R)-(-)-Thiazolidine-4-carboxylic acid

Hazard: Toxic

Formula: C₄H₇NO₂S
Melting Pt: 191-200°C
MDL Number: MFCD00005212
CAS Number: 34592-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR