2-Chloro-4-fluorophenylhydrazine
Supplier: Matrix Scientific
037412-5G
037412-25G
101843-312EA
41.54
USD
101843-312
101843-314
2-Chloro-4-fluorophenylhydrazine
2-Chloro-4-fluorophenylhydrazine
Hazard: Irritant
Formula:
C₆H₆ClFN₂ |
MDL Number:
MFCD03094567 CAS Number: 119452-65-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...