DL-Ornithine hydrochloride

Supplier: Matrix Scientific

Synonyms: H-DL-Orn-OH·HCl, 2,5-Diaminopentanoic acid hydrochloride, DL-Ornithine monohydrochloride

037401-25G 037401-100G
101843-288EA 110.77 USD
101843-288 101843-290
DL-Ornithine hydrochloride
DL-Ornithine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
Melting Pt: ∼ 225 °C (dec.)
MDL Number: MFCD00065398
CAS Number: 1069-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR