1-(Chloromethyl)-2-(trifluoromethoxy)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethoxy)benzyl chloride

037400-5G 037400-25G
101843-284EA 30.77 USD
101843-284 101843-286
1-(Chloromethyl)-2-(trifluoromethoxy)benzene ≥97%
1-(Chloromethyl)-2-(trifluoromethoxy)benzene

Hazard: Irritant

Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
MDL Number: MFCD00221456
CAS Number: 116827-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR