Tetrahydropyran-4-methanol ≥95%

Supplier: Matrix Scientific

037091-5G 037091-25G
101842-654EA 100 USD
101842-654 102833-048
Tetrahydropyran-4-methanol ≥95%
Tetrahydropyran-4-methanol

Hazard: Irritant/Stored under Argon

Formula: C₆H₁₂O₂
MDL Number: MFCD00457804
CAS Number: 14774-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR