5-Bromo-2-methoxyphenol ≥98%

Supplier: Matrix Scientific

035780-5G 035780-25G 035780-100G
101840-048EA 15.38 USD
101840-048 101840-050 101840-052
5-Bromo-2-methoxyphenol ≥98%
5-Bromo-2-methoxyphenol

Hazard: Irritant

Formula: C₇H₇BrO₂
MW: 203.0300 g/mol
MDL Number: MFCD00673137
CAS Number: 37942-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR