1-(7-Bromo-4-methyl-3,4-dihydropyrido[2,3-b]pyrazin-1(2H)-yl)-2,2-dimethylpropan-1-one

Supplier: Matrix Scientific

035779-100MG 035779-500MG
101840-044EA 364.62 USD
101840-044 101840-046
1-(7-Bromo-4-methyl-3,4-dihydropyrido[2,3-b]pyrazin-1(2H)-yl)-2,2-dimethylpropan-1-one
1-(7-Bromo-4-methyl-3,4-dihydropyrido[2,3-b]pyrazin-1(2H)-yl)-2,2-dimethylpropan-1-one

Hazard: Irritant

Formula: C₁₃H₁₈BrN₃O
MDL Number: MFCD11857751
CAS Number: 1142192-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR